Assay Details
Target Gene Details
Entrez Gene ID: | 10200 |
Gene Name: | M-phase phosphoprotein 6 |
Gene Aliases: |
MPP, MPP-6, MPP6 |
Location: |
Chr.16:82148162-82170224 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPHOSPH6 | NM_005792.2 | 5 | 984 | NP_005783.2 |
| XM_011522808.2 | 6 | 1264 | XP_011521110.1 | |
| XM_017022852.1 | 5 | 1098 | XP_016878341.1 | |
| AI831131.1 | 1 | 118 | ||
| AK222667.1 | 5 | 984 | BAD96387.1 | |
| BC005242.1 | 5 | 955 | AAH05242.1 | |
| BC011020.1 | 5 | 961 | AAH11020.1 | |
| BC031017.1 | 5 | 956 | AAH31017.1 | |
| BU622913.1 | ||||
| X98263.1 | 5 | 967 | CAA66916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3050n100 | Chr.16:82131709 - 82181783 on Build GRCh38 | Loss |
|
| dgv5252n54 | Chr.16:82131578 - 82151715 on Build GRCh38 | Loss |
|
| dgv5251n54 | Chr.16:82102961 - 82183017 on Build GRCh38 | Loss |
|
| esv3639349 | Chr.16:82089507 - 82148922 on Build GRCh38 | Gain |
|
| esv3639354 | Chr.16:82134351 - 82201089 on Build GRCh38 | Gain |
|
| nsv573367 | Chr.16:82106956 - 83007241 on Build GRCh38 | Gain |
|
| nsv1061288 | Chr.16:82146152 - 83636829 on Build GRCh38 | Gain+Loss |
|
| dgv3051n100 | Chr.16:82138730 - 83654472 on Build GRCh38 | Gain |
|
| nsv525467 | Chr.16:82121014 - 82189880 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map