Assay Details
Target Gene Details
Entrez Gene ID: | 144983 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A1-like 2 |
Gene Aliases: |
- |
Location: |
Chr.13:52617470-52643784 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPA1L2 | NM_001011724.1 | 1 | 94 | NP_001011724.1 |
| NM_001011725.1 | 1 | 94 | NP_001011725.1 | |
| AK123081.1 | ||||
| AK123159.1 | 1 | 50 | ||
| BQ425201.1 | 1 | 62 | ||
| BX477483.1 | 1 | 96 | ||
| CD523364.1 | 1 | 65 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3632103 | Chr.13:52601435 - 52636346 on Build GRCh38 | Loss |
|
| nsv976119 | Chr.13:52601894 - 52644361 on Build GRCh38 | Duplication |
|
| nsv1044949 | Chr.13:52497005 - 52715291 on Build GRCh38 | Gain |
|
| esv3892344 | Chr.13:52454575 - 52629536 on Build GRCh38 | Loss |
|
| dgv1653n100 | Chr.13:52564997 - 52728939 on Build GRCh38 | Gain |
|
| nsv820041 | Chr.13:52487203 - 52706453 on Build GRCh38 | Gain |
|
| esv3892345 | Chr.13:52564080 - 52625702 on Build GRCh38 | Gain |
|
| nsv819201 | Chr.13:52563355 - 52666195 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map