Assay Details
Target Gene Details
Entrez Gene ID: | 92345 |
Gene Name: | nuclear assembly factor 1 ribonucleoprotein |
Gene Aliases: |
- |
Location: |
Chr.4:163109133-163170388 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAF1 | NM_001128931.1 | 5 | 977 | NP_001122403.1 |
| NM_138386.2 | 5 | 977 | NP_612395.2 | |
| XM_011532410.2 | 5 | 4444 | XP_011530712.1 | |
| XM_011532411.2 | 6 | 1051 | XP_011530713.1 | |
| AK097352.1 | 5 | 887 | ||
| AK308601.1 | 5 | 946 | ||
| BC008207.1 | 5 | 977 | AAH08207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015505 | Chr.4:161989825 - 163516137 on Build GRCh38 | Gain |
|
| nsv1030184 | Chr.4:162402427 - 164455529 on Build GRCh38 | Loss |
|
| esv3894064 | Chr.4:162963457 - 163303980 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map