Assay Details
Target Gene Details
Entrez Gene ID: | 84902 |
Gene Name: | centrosomal protein 89 |
Gene Aliases: |
CCDC123, CEP123 |
Location: |
Chr.19:32875925-32972029 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP89 | NM_032816.4 | NP_116205.3 | ||
| XM_005259344.3 | XP_005259401.1 | |||
| XM_011527425.2 | XP_011525727.1 | |||
| XM_017027398.1 | XP_016882887.1 | |||
| AK027546.1 | BAB55190.1 | |||
| AL832158.1 | ||||
| BC136328.1 | ||||
| BE208545.1 | ||||
| BU196203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062926 | Chr.19:32828718 - 32904288 on Build GRCh38 | Loss |
|
| nsv833807 | Chr.19:32889838 - 33077690 on Build GRCh38 | Loss |
|
| nsv1060014 | Chr.19:32854920 - 33011720 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map