Assay Details
Target Gene Details
Entrez Gene ID: | 79184 |
Gene Name: | BRCA1/BRCA2-containing complex subunit 3 |
Gene Aliases: |
BRCC36, C6.1A, CXorf53 |
Location: |
Chr.X:155071403-155126766 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRCC3 | NM_001018055.2 | NP_001018065.1 | ||
| NM_001242640.1 | NP_001229569.1 | |||
| NM_024332.3 | NP_077308.1 | |||
| XM_005274751.4 | XP_005274808.1 | |||
| XM_017029838.1 | XP_016885327.1 | |||
| AK298886.1 | ||||
| AK299194.1 | ||||
| AK310286.1 | ||||
| AK313544.1 | ||||
| AY438030.1 | AAR30498.1 | |||
| BC002999.1 | AAH02999.1 | |||
| BC006540.2 | AAH06540.1 | |||
| BP304352.1 | ||||
| S68015.1 | AAB29005.2 | |||
| X64643.1 | CAA45917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| esv29815 | Chr.X:155071577 - 155217315 on Build GRCh38 | Gain |
|
| esv29972 | Chr.X:154781072 - 155699751 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map