Assay Details
Target Gene Details
Entrez Gene ID: | 10776 |
Gene Name: | cAMP regulated phosphoprotein 19 |
Gene Aliases: |
ARPP-16, ARPP-19, ARPP16, ENSAL |
Location: |
Chr.15:52547045-52569446 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPP19 | NM_001306191.1 | 6 | 1313 | NP_001293120.1 |
| NM_001306195.1 | 4 | 1131 | NP_001293124.1 | |
| NM_001306196.1 | 4 | 1312 | NP_001293125.1 | |
| NM_006628.5 | 3 | 1255 | NP_006619.1 | |
| XM_017021868.1 | 4 | 1076 | XP_016877357.1 | |
| XM_017021869.1 | 4 | 1571 | XP_016877358.1 | |
| XM_017021870.1 | 3 | 1065 | XP_016877359.1 | |
| XM_017021871.1 | 3 | 1514 | XP_016877360.1 | |
| AF084555.1 | 3 | 1139 | AAD52044.1 | |
| AK123616.1 | 3 | 1514 | ||
| AL833077.2 | 4 | 1078 | CAD89929.1 | |
| BC003418.1 | 3 | 1139 | AAH03418.1 | |
| BC100015.1 | 4 | 1064 | AAI00016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833006 | Chr.15:52533757 - 52748265 on Build GRCh38 | Gain |
|
| nsv1047775 | Chr.15:52529853 - 52620031 on Build GRCh38 | Gain |
|
| esv1001541 | Chr.15:52547145 - 52551659 on Build GRCh38 | Gain |
|
| nsv471245 | Chr.15:52315065 - 52894968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map