Assay Details
Target Gene Details
Entrez Gene ID: | 353288 |
Gene Name: | keratin 26 |
Gene Aliases: |
CK26, K25, K25IRS2, K26, KRT25B |
Location: |
Chr.17:40766238-40772159 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT26 | NM_181539.4 | 1 | 228 | NP_853517.2 |
| AJ564205.2 | 1 | 228 | CAD91905.1 | |
| BC132951.1 | 1 | 219 | ||
| BC136896.1 | 1 | 219 |
More Information
Set Membership: |
|

Genomic Map