Assay Details
Target Gene Details
Entrez Gene ID: | 844 |
Gene Name: | calsequestrin 1 |
Gene Aliases: |
CASQ, PDIB1, VMCQA |
Location: |
Chr.1:160190495-160201886 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASQ1 | NM_001231.4 | 11 | 1650 | NP_001222.3 |
| AA197256.1 | 1 | 344 | ||
| AI141895.1 | 1 | 317 | ||
| AK300713.1 | ||||
| BC022289.1 | 11 | 1536 | AAH22289.1 | |
| S73775.1 | 11 | 1540 | AAB32063.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs74123289] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map