Assay Details
Target Gene Details
Entrez Gene ID: | 5993 |
Gene Name: | regulatory factor X5 |
Gene Aliases: |
- |
Location: |
Chr.1:151340640-151347319 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX5 | NM_000449.3 | 11 | 3344 | NP_000440.1 |
| NM_001025603.1 | 11 | 3337 | NP_001020774.1 | |
| XM_005245405.1 | 11 | 3399 | XP_005245462.1 | |
| XM_005245406.3 | 11 | 3356 | XP_005245463.1 | |
| XM_011509847.1 | 11 | 3403 | XP_011508149.1 | |
| XM_011509848.1 | 11 | 3406 | XP_011508150.1 | |
| XM_011509849.1 | 11 | 3339 | XP_011508151.1 | |
| XM_011509850.1 | 11 | 3466 | XP_011508152.1 | |
| XM_017001999.1 | 11 | 3467 | XP_016857488.1 | |
| XM_017002000.1 | 11 | 3498 | XP_016857489.1 | |
| AL050135.1 | 3 | 2579 | CAB43285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| esv34461 | Chr.1:151268066 - 151439159 on Build GRCh38 | Gain |
|
| nsv831581 | Chr.1:151316675 - 151506129 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map