Assay Details
Target Gene Details
Entrez Gene ID: | 26058 |
Gene Name: | GRB10 interacting GYF protein 2 |
Gene Aliases: |
GYF2, PARK11, PERQ2, PERQ3, TNRC15 |
Location: |
Chr.2:232697305-232860577 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIGYF2 | NM_001103146.1 | NP_001096616.1 | ||
| NM_001103147.1 | NP_001096617.1 | |||
| NM_001103148.1 | NP_001096618.1 | |||
| NM_015575.3 | NP_056390.2 | |||
| NR_103492.1 | ||||
| AB014542.2 | ||||
| AK001739.1 | BAA91873.1 | |||
| AK297162.1 | ||||
| AK299418.1 | ||||
| AK302998.1 | ||||
| AY176045.1 | AAO46889.1 | |||
| BC136251.1 | ||||
| BC146775.1 | ||||
| BX537885.1 | CAD97881.1 | |||
| BX538172.1 | ||||
| BX538321.1 | CAD98095.1 | |||
| BX647505.1 |
Target Gene Details
Entrez Gene ID: | 3769 |
Gene Name: | potassium voltage-gated channel subfamily J member 13 |
Gene Aliases: |
KIR1.4, KIR7.1, LCA16, SVD |
Location: |
Chr.2:232765802-232776565 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNJ13 | NM_001172416.1 | 3 | 1989 | NP_001165887.1 |
| NM_001172417.1 | 3 | 1992 | NP_001165888.1 | |
| NM_002242.4 | 3 | 2225 | NP_002233.2 | |
| AJ007557.1 | 3 | 2204 | CAA07552.1 | |
| BC037290.2 | 3 | 2225 | AAH37290.1 |
More Information
Set Membership: |
|

Genomic Map