Assay Details
Target Gene Details
Entrez Gene ID: | 64231 |
Gene Name: | membrane spanning 4-domains A6A |
Gene Aliases: |
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090 |
Location: |
Chr.11:60171607-60184666 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A6A | NM_001247999.1 | 6 | 1044 | NP_001234928.1 |
| NM_022349.3 | 5 | 855 | NP_071744.2 | |
| NM_152851.2 | NP_690590.1 | |||
| NM_152852.2 | 6 | 980 | NP_690591.1 | |
| XM_005274177.2 | 6 | 1039 | XP_005274234.1 | |
| XM_006718658.2 | 6 | 914 | XP_006718721.1 | |
| XM_006718660.2 | 5 | 779 | XP_006718723.1 | |
| XM_006718661.2 | 5 | 779 | XP_006718724.1 | |
| XM_017018123.1 | 4 | 515 | XP_016873612.1 | |
| XM_017018124.1 | 4 | 515 | XP_016873613.1 | |
| AB013104.1 | BAB18740.1 | |||
| AF142409.1 | 5 | 583 | AAG27920.1 | |
| AF212240.1 | 5 | 724 | AAG41780.1 | |
| AF237908.1 | 5 | 725 | AAK37417.1 | |
| AF253977.1 | 5 | 725 | AAG44626.1 | |
| AF261136.1 | 5 | 725 | AAG44672.1 | |
| AF286866.1 | 4 | 503 | AAK37994.1 | |
| AF350502.1 | 5 | 572 | AAL56222.1 | |
| AF350503.1 | 5 | 572 | AAL56223.1 | |
| AF354930.1 | 5 | 725 | AAL07357.1 | |
| AK291870.1 | 6 | 980 | ||
| AK300725.1 | 5 | 589 | ||
| AK301387.1 | 4 | 415 | ||
| BC022854.1 | 5 | 594 | AAH22854.1 | |
| BC108728.1 | 5 | 628 | AAI08729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832174 | Chr.11:60141794 - 60306435 on Build GRCh38 | Gain |
|
| nsv555150 | Chr.11:60046814 - 60207927 on Build GRCh38 | Gain |
|
| nsv524137 | Chr.11:60169506 - 60179779 on Build GRCh38 | Loss |
|
| esv33843 | Chr.11:60140658 - 60177861 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map