Assay Details
Target Gene Details
Entrez Gene ID: | 25791 |
Gene Name: | neuronal guanine nucleotide exchange factor |
Gene Aliases: |
ARHGEF27, EPHEXIN |
Location: |
Chr.2:232878686-233013241 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NGEF | NM_001114090.1 | 10 | 1600 | NP_001107562.1 |
| NM_019850.2 | 12 | 1972 | NP_062824.2 | |
| XM_011510923.2 | 12 | 1938 | XP_011509225.1 | |
| XM_011510924.1 | 8 | 971 | XP_011509226.1 | |
| XM_011510925.1 | 8 | 975 | XP_011509227.1 | |
| AJ238899.1 | CAC00686.1 | |||
| AK293203.1 | 8 | 972 | ||
| AK297390.1 | 10 | 1600 | ||
| AK298315.1 | 12 | 1924 | ||
| AK300953.1 | 13 | 1876 | ||
| BC031573.1 | 12 | 1959 | AAH31573.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4188n100 | Chr.2:232868642 - 232895809 on Build GRCh38 | Gain |
|
| nsv953203 | Chr.2:232867891 - 232898190 on Build GRCh38 | Deletion |
|
| nsv1009825 | Chr.2:232844856 - 232898760 on Build GRCh38 | Gain |
|
| nsv834570 | Chr.2:232774450 - 232940357 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map