Assay Details
Target Gene Details
Entrez Gene ID: | 1382 |
Gene Name: | cellular retinoic acid binding protein 2 |
Gene Aliases: |
CRABP-II, RBP6 |
Location: |
Chr.1:156699606-156712900 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRABP2 | NM_001199723.1 | 5 | 761 | NP_001186652.1 |
| NM_001878.3 | 4 | 774 | NP_001869.1 | |
| XM_017000343.1 | 7 | 6188 | XP_016855832.1 | |
| AB593017.1 | 4 | 690 | ||
| BC001109.2 | 4 | 676 | AAH01109.1 | |
| M68867.1 | 4 | 651 | AAA52068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| nsv523181 | Chr.1:156580462 - 156706761 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map