Assay Details
Target Gene Details
Entrez Gene ID: | 3624 |
Gene Name: | inhibin beta A subunit |
Gene Aliases: |
EDF, FRP |
Location: |
Chr.7:41685101-41710532 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INHBA | NM_002192.2 | 3 | 918 | NP_002183.1 |
| XM_017012174.1 | 5 | 2810 | XP_016867663.1 | |
| XM_017012175.1 | 3 | 2494 | XP_016867664.1 | |
| XM_017012176.1 | 5 | 2678 | XP_016867665.1 | |
| AF218018.1 | 3 | 877 | AAG17260.1 | |
| AK222742.1 | 2 | 747 | BAD96462.1 | |
| AK290584.1 | 3 | 881 | ||
| BC007858.2 | 3 | 888 | AAH07858.1 | |
| BX648811.1 | 2 | 1659 | CAI46003.1 | |
| J03634.1 | 2 | 756 | AAA35787.1 | |
| M13436.1 | 2 | 907 | AAA59168.1 |
More Information
Set Membership: |
|

Genomic Map