Assay Details
Target Gene Details
Entrez Gene ID: | 113612 |
Gene Name: | cytochrome P450 family 2 subfamily U member 1 |
Gene Aliases: |
P450TEC, SPG49, SPG56 |
Location: |
Chr.4:107931549-107953461 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP2U1 | NM_183075.2 | NP_898898.1 | ||
| XM_005262717.2 | XP_005262774.1 | |||
| XM_005262720.2 | XP_005262777.1 | |||
| AK290002.1 | ||||
| AK303284.1 | ||||
| AK304724.1 | ||||
| AY343323.1 | AAQ21380.1 | |||
| BC012027.1 | AAH12027.1 | |||
| BC132767.1 | ||||
| BC136483.1 | ||||
| BX354124.2 | ||||
| BX437421.2 |
Target Gene Details
Entrez Gene ID: | 101929595 |
Gene Name: | uncharacterized LOC101929595 |
Gene Aliases: |
- |
Location: |
Chr.4:107863479-107932119 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929595 | NR_125929.1 | |||
| AK123292.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161125 | Chr.4:107930839 - 107944992 on Build GRCh38 | Deletion |
|
| nsv461611 | Chr.4:107897601 - 107966289 on Build GRCh38 | Loss |
|
| nsv4457 | Chr.4:107910943 - 107941846 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map