Assay Details
Target Gene Details
Entrez Gene ID: | 4625 |
Gene Name: | myosin, heavy chain 7, cardiac muscle, beta |
Gene Aliases: |
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM |
Location: |
Chr.14:23412738-23435686 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYH7 | NM_000257.3 | NP_000248.2 | ||
| XM_017021340.1 | XP_016876829.1 | |||
| AK307590.1 | ||||
| BC112171.1 | AAI12172.1 | |||
| BC112173.1 | AAI12174.1 | |||
| BF834726.1 | ||||
| DA823104.1 | ||||
| EF560725.1 | ||||
| EU747717.1 | ||||
| M58018.1 | AAA62830.1 |
More Information
Set Membership: |
|

Genomic Map