Assay Details
Target Gene Details
Entrez Gene ID: | 54885 |
Gene Name: | TBC1 domain family member 8B |
Gene Aliases: |
GRAMD8B |
Location: |
Chr.X:106802689-106876150 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D8B | NM_017752.2 | 7 | 1271 | NP_060222.2 |
| NM_198881.1 | 7 | 1271 | NP_942582.1 | |
| XM_006724669.1 | 7 | 1271 | XP_006724732.1 | |
| XM_006724670.3 | 7 | 1085 | XP_006724733.1 | |
| AB449890.1 | 7 | 1106 | ||
| AB449891.1 | 7 | 1106 | ||
| AK123957.1 | 7 | 1271 | BAC85735.1 | |
| BC122564.1 | 7 | 1270 | ||
| BX648230.1 | 7 | 1255 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752312 | Chr.X:105705935 - 107123872 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map