Assay Details
Target Gene Details
Entrez Gene ID: | 93487 |
Gene Name: | mitogen-activated protein kinase 1 interacting protein 1 like |
Gene Aliases: |
C14orf32, MISS, c14_5346 |
Location: |
Chr.14:55051608-55070194 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK1IP1L | NM_144578.3 | 4 | 2076 | NP_653179.1 |
| XM_011537362.2 | 4 | 2920 | XP_011535664.2 | |
| AK025580.1 | ||||
| AL834285.1 | 4 | 2059 | CAD38959.2 | |
| U90909.1 | 1 | 1071 |
More Information
Additional Information:
For this assay, SNP(s) [rs115758572] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map