Assay Details
Target Gene Details
Entrez Gene ID: | 102 |
Gene Name: | ADAM metallopeptidase domain 10 |
Gene Aliases: |
AD10, AD18, CD156c, CDw156, HsT18717, MADM, RAK, kuz |
Location: |
Chr.15:58595204-58749978 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM10 | NM_001110.3 | 14 | 2373 | NP_001101.1 |
| NM_001320570.1 | 13 | 2280 | NP_001307499.1 | |
| XM_017021861.1 | 15 | 2428 | XP_016877350.1 | |
| AF009615.1 | 14 | 2398 | AAC51766.1 | |
| AK023460.1 | 2 | 1633 | ||
| AK300472.1 | 8 | 1122 | ||
| BC126253.1 | 14 | 2203 | ||
| Z48579.1 | 12 | 1758 | CAA88463.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1551 | Chr.15:58599440 - 58644381 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map