Assay Details
Target Gene Details
Entrez Gene ID: | 3433 |
Gene Name: | interferon induced protein with tetratricopeptide repeats 2 |
Gene Aliases: |
G10P2, GARG-39, IFI-54, IFI-54K, IFI54, IFIT-2, ISG-54 K, ISG-54K, ISG54, P54, cig42 |
Location: |
Chr.10:89301949-89309276 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFIT2 | NM_001547.4 | 2 | 2461 | NP_001538.4 |
| BC032839.2 | 2 | 2364 | AAH32839.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34085 | Chr.10:89306192 - 89414418 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map