Assay Details
Target Gene Details
Entrez Gene ID: | 57097 |
Gene Name: | poly(ADP-ribose) polymerase family member 11 |
Gene Aliases: |
ARTD11, C12orf6, MIB006 |
Location: |
Chr.12:3808861-3873448 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP11 | NM_001286521.1 | 7 | 3432 | NP_001273450.1 |
| NM_001286522.1 | 8 | 3554 | NP_001273451.1 | |
| NM_020367.5 | 8 | 3553 | NP_065100.2 | |
| NR_104461.1 | 9 | 3612 | ||
| AK096373.1 | 1 | 1121 | ||
| BX640960.1 | 7 | 3356 | CAE45979.1 | |
| BX647487.1 | 8 | 3472 | ||
| CR749294.1 | 8 | 3460 | CAH18149.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv468981 | Chr.12:3576597 - 3989342 on Build GRCh38 | Gain |
|
| nsv483060 | Chr.12:3190835 - 5290834 on Build GRCh38 | Loss |
|
| nsv832313 | Chr.12:3689562 - 3906773 on Build GRCh38 | Loss |
|
| nsv1036828 | Chr.12:3607219 - 3846326 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map