Assay Details
Target Gene Details
Entrez Gene ID: | 995 |
Gene Name: | cell division cycle 25C |
Gene Aliases: |
CDC25, PPP1R60 |
Location: |
Chr.5:138285265-138338355 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC25C | NM_001287582.1 | 7 | 837 | NP_001274511.1 |
| NM_001287583.1 | 7 | 954 | NP_001274512.1 | |
| NM_001318098.1 | 7 | 958 | NP_001305027.1 | |
| NM_001790.4 | 7 | 841 | NP_001781.2 | |
| NM_022809.3 | 4 | 622 | NP_073720.1 | |
| XM_005272145.3 | 7 | 954 | XP_005272202.1 | |
| XM_006714739.3 | 6 | 552 | XP_006714802.1 | |
| XM_011543759.2 | 8 | 625 | XP_011542061.1 | |
| XM_011543760.2 | 7 | 954 | XP_011542062.1 | |
| XM_011543761.2 | 4 | 735 | XP_011542063.1 | |
| XM_011543762.1 | 4 | 535 | XP_011542064.1 | |
| XM_011543763.1 | 5 | 250 | XP_011542065.1 | |
| XM_011543764.1 | 4 | 303 | XP_011542066.1 | |
| XM_017010101.1 | 7 | 832 | XP_016865590.1 | |
| AF277723.1 | 5 | 337 | AAG41885.1 | |
| AF277724.1 | 4 | 303 | AAG41886.1 | |
| AF277725.1 | 4 | 298 | AAG41887.1 | |
| AF277726.1 | 3 | 208 | AAG41888.1 | |
| AF312681.1 | 2 | 84 | AAL26835.1 | |
| AJ304504.1 | 3 | 301 | CAC19192.1 | |
| AK097710.1 | 7 | 954 | ||
| AK301828.1 | 8 | 904 | ||
| AU125025.1 | ||||
| BC019089.2 | 7 | 731 | AAH19089.1 | |
| M34065.1 | 7 | 730 | AAA35666.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033604 | Chr.5:138291351 - 138404936 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map