Assay Details
Target Gene Details
Entrez Gene ID: | 23157 |
Gene Name: | septin 6 |
Gene Aliases: |
SEP2, SEPT2 |
Location: |
Chr.X:119615724-119693370 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEPT6 | NM_015129.5 | 10 | 3275 | NP_055944.2 |
| NM_145799.3 | 11 | 4585 | NP_665798.1 | |
| NM_145800.3 | NP_665799.1 | |||
| NM_145802.3 | NP_665801.1 | |||
| XM_005262400.3 | 11 | 4410 | XP_005262457.1 | |
| XM_006724748.2 | 10 | 4348 | XP_006724811.1 | |
| XM_006724750.2 | 11 | 3128 | XP_006724813.1 | |
| XM_011531317.2 | XP_011529619.1 | |||
| XM_011531318.2 | 11 | 3145 | XP_011529620.1 | |
| AF397023.1 | AAN76547.1 | |||
| AF403060.1 | AAK98549.1 | |||
| AF403061.1 | AAK98550.1 | |||
| AF403062.1 | AAK98551.1 | |||
| AK026589.1 | ||||
| AK290977.1 | ||||
| AL833677.1 | 1 | 2888 | ||
| AY034177.1 | 11 | 4575 | AAK61492.1 | |
| AY679521.1 | AAT80339.1 | |||
| BC011922.2 | AAH11922.3 |
More Information
Set Membership: |
|

Genomic Map