Assay Details
Target Gene Details
Entrez Gene ID: | 84807 |
Gene Name: | NFKB inhibitor delta |
Gene Aliases: |
IkappaBNS, TA-NFKBH |
Location: |
Chr.19:35887630-35902301 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Intron 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv51n68 | Chr.19:35841011 - 36016692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map