Assay Details
Target Gene Details
Entrez Gene ID: | 54985 |
Gene Name: | host cell factor C1 regulator 1 |
Gene Aliases: |
HPIP |
Location: |
Chr.16:3022620-3024286 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCFC1R1 | NM_001002017.2 | NP_001002017.1 | ||
| NM_001002018.2 | NP_001002018.1 | |||
| NM_001288665.1 | NP_001275594.1 | |||
| NM_001288666.1 | NP_001275595.1 | |||
| NM_001288667.1 | NP_001275596.1 | |||
| NM_001288668.1 | NP_001275597.1 | |||
| NM_001308070.1 | NP_001294999.1 | |||
| NM_017885.3 | NP_060355.1 | |||
| XM_011522559.2 | XP_011520861.1 | |||
| XM_017023384.1 | XP_016878873.1 | |||
| AK000575.1 | BAA91265.1 | |||
| AK293768.1 | ||||
| AY116892.1 | AAM49798.1 | |||
| BC064026.1 | AAH64026.1 | |||
| BC080178.1 | AAH80178.1 | |||
| BM790570.1 | ||||
| BU623445.1 | ||||
| BU633901.1 | ||||
| CN427571.1 | ||||
| CR457245.1 | CAG33526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637645 | Chr.16:3021600 - 3057596 on Build GRCh38 | Gain |
|
| nsv509588 | Chr.16:2919880 - 3043776 on Build GRCh38 | Insertion |
|
| nsv833125 | Chr.16:3017124 - 3131817 on Build GRCh38 | Loss |
|
| nsv952906 | Chr.16:2868600 - 3028099 on Build GRCh38 | Deletion |
|

Genomic Map