Assay Details
Target Gene Details
Entrez Gene ID: | 5743 |
Gene Name: | prostaglandin-endoperoxide synthase 2 |
Gene Aliases: |
COX-2, COX2, GRIPGHS, PGG/HS, PGHS-2, PHS-2, hCox-2 |
Location: |
Chr.1:186671812-186680427 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGS2 | NM_000963.3 | 7 | 1020 | NP_000954.1 |
| AJ634912.1 | 7 | 773 | CAG25548.1 | |
| AK292167.1 | 7 | 1008 | ||
| AY151286.1 | 7 | 980 | AAN52932.1 | |
| AY462100.1 | 7 | 883 | AAR23927.1 | |
| BC013734.1 | 7 | 1003 | AAH13734.1 | |
| L15326.1 | 7 | 975 | AAA35803.1 | |
| M90100.1 | 7 | 980 | AAA58433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548403 | Chr.1:186673297 - 186731827 on Build GRCh38 | Loss |
|
| esv2762854 | Chr.1:186577969 - 186878442 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map