Assay Details
Target Gene Details
Entrez Gene ID: | 403150 |
Gene Name: | uncharacterized protein FLJ31356 |
Gene Aliases: |
- |
Location: |
Chr.2:28384409-28394672 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ31356 | NR_103831.1 | |||
| AK055918.1 |
Target Gene Details
Entrez Gene ID: | 2355 |
Gene Name: | FOS like 2, AP-1 transcription factor subunit |
Gene Aliases: |
FRA2 |
Location: |
Chr.2:28392759-28417312 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOSL2 | NM_005253.3 | 1 | 552 | NP_005244.1 |
| XM_005264231.3 | 1 | 705 | XP_005264288.1 | |
| XM_006711976.2 | 1 | 705 | XP_006712039.1 | |
| AK055579.1 | 1 | 606 | ||
| BC022791.1 | AAH22791.1 | |||
| BM023443.1 | ||||
| BQ574214.1 | 1 | 122 | ||
| BX647822.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828664 | Chr.2:28391689 - 28394040 on Build GRCh38 | Gain |
|
| nsv1006830 | Chr.2:28331908 - 28419880 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map