Assay Details
Target Gene Details
Entrez Gene ID: | 64429 |
Gene Name: | zinc finger DHHC-type containing 6 |
Gene Aliases: |
DHHC-6, ZNF376 |
Location: |
Chr.10:112424411-112448335 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss |
|
| nsv831991 | Chr.10:112413098 - 112582139 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map