Assay Details
Target Gene Details
Entrez Gene ID: | 79365 |
Gene Name: | basic helix-loop-helix family member e41 |
Gene Aliases: |
BHLHB3, DEC2, SHARP1, hDEC2 |
Location: |
Chr.12:26120026-26125070 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BHLHE41 | NM_030762.2 | NP_110389.1 | ||
| AB044088.1 | BAB21502.1 | |||
| AK303677.1 | ||||
| AY665467.1 | AAV54602.1 | |||
| BC025968.1 | AAH25968.1 |
Target Gene Details
Entrez Gene ID: | 8082 |
Gene Name: | sarcospan |
Gene Aliases: |
DAGA5, KRAG, NSPN, SPN1, SPN2 |
Location: |
Chr.12:26122024-26234777 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSPN | XM_011520853.2 | XP_011519155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581126 | Chr.12:26119891 - 26132037 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map