Assay Details
Target Gene Details
Entrez Gene ID: | 79843 |
Gene Name: | family with sequence similarity 124 member B |
Gene Aliases: |
- |
Location: |
Chr.2:224378698-224401994 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM124B | NM_001122779.1 | 2 | 1856 | NP_001116251.1 |
| NM_024785.2 | 3 | 1981 | NP_079061.2 | |
| AK026399.1 | 2 | 1896 | BAB15471.1 | |
| AK075126.1 | 3 | 1484 | ||
| BC025754.1 | 3 | 1981 | AAH25754.1 |
More Information
Set Membership: |
|

Genomic Map