Assay Details
Target Gene Details
Entrez Gene ID: | 55609 |
Gene Name: | zinc finger protein 280C |
Gene Aliases: |
SUHW3, ZNF633 |
Location: |
Chr.X:130202699-130268948 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF280C | NM_017666.4 | 19 | 2442 | NP_060136.1 |
| XM_006724765.3 | 18 | 2246 | XP_006724828.1 | |
| AK290373.1 | 19 | 2393 | ||
| AL834333.1 | 19 | 2373 | CAD39001.1 | |
| BC051728.1 | 19 | 2393 | AAH51728.1 | |
| DA441065.1 | 6 | 466 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528290 | Chr.X:129635682 - 130429852 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map