Assay Details
Target Gene Details
Entrez Gene ID: | 4635 |
Gene Name: | myosin light chain 4 |
Gene Aliases: |
ALC1, AMLC, GT1, PRO1957 |
Location: |
Chr.17:47209017-47227650 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYL4 | NM_001002841.1 | NP_001002841.1 | ||
| NM_002476.2 | NP_002467.1 | |||
| XM_005257391.4 | XP_005257448.1 | |||
| XM_011524839.2 | XP_011523141.2 | |||
| XM_017024683.1 | XP_016880172.1 | |||
| XM_017024684.1 | XP_016880173.1 | |||
| BC030228.1 | AAH30228.1 | |||
| BU658678.1 | ||||
| M24121.1 | AAA59891.1 | |||
| M36172.1 | AAA36319.1 | |||
| X13955.1 | CAA32137.1 |
More Information
Set Membership: |
|

Genomic Map