Assay Details
Target Gene Details
Entrez Gene ID: | 4121 |
Gene Name: | mannosidase alpha class 1A member 1 |
Gene Aliases: |
HUMM3, HUMM9, MAN9 |
Location: |
Chr.6:119177201-119350619 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAN1A1 | NM_005907.3 | NP_005898.2 | ||
| XM_005266986.4 | XP_005267043.1 | |||
| XM_011535833.2 | XP_011534135.1 | |||
| XM_011535834.2 | XP_011534136.1 | |||
| AK025599.1 | ||||
| AK307383.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034992 | Chr.6:119191492 - 119219730 on Build GRCh38 | Loss |
|
| nsv1016699 | Chr.6:119139354 - 120348940 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map