Assay Details
Target Gene Details
Entrez Gene ID: | 2121 |
Gene Name: | EvC ciliary complex subunit 1 |
Gene Aliases: |
DWF-1, EVC1, EVCL |
Location: |
Chr.4:5711197-5829043 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EVC | NM_001306090.1 | 9 | 1358 | NP_001293019.1 |
| NM_001306092.1 | 9 | 1358 | NP_001293021.1 | |
| NM_153717.2 | 9 | 1358 | NP_714928.1 | |
| XM_006713865.3 | 9 | 1355 | XP_006713928.1 | |
| XM_006713866.3 | 9 | 1355 | XP_006713929.1 | |
| XM_011513419.2 | 9 | 1352 | XP_011511721.1 | |
| XM_017007883.1 | 9 | 1352 | XP_016863372.1 | |
| AF216184.1 | 9 | 1358 | AAF37216.1 | |
| AF216185.1 | 9 | 1358 | AAF37217.1 | |
| AF239742.1 | 9 | 1174 | AAF44682.1 | |
| AK025394.1 | 4 | 487 | ||
| BC085608.1 | 9 | 1340 | AAH85608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv461182 | Chr.4:5733576 - 5780257 on Build GRCh38 | Loss |
|
| nsv1000365 | Chr.4:5747389 - 5769007 on Build GRCh38 | Gain |
|
| nsv829844 | Chr.4:5719377 - 5925385 on Build GRCh38 | Gain |
|
| nsv593480 | Chr.4:4961200 - 5760434 on Build GRCh38 | Loss |
|
| nsv522904 | Chr.4:5752388 - 5757156 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map