Assay Details
Target Gene Details
Entrez Gene ID: | 219493 |
Gene Name: | olfactory receptor family 5 subfamily AR member 1 (gene/pseudogene) |
Gene Aliases: |
OR11-209 |
Location: |
Chr.11:56663686-56664618 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR5AR1 | NM_001004730.1 | 1 | 736 | NP_001004730.1 |
| BC140745.1 | 1 | 760 |
More Information
Set Membership: |
|

Genomic Map