Assay Details
Target Gene Details
Entrez Gene ID: | 50999 |
Gene Name: | transmembrane p24 trafficking protein 5 |
Gene Aliases: |
CGI-100, p24g2, p28 |
Location: |
Chr.1:93149742-93180689 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED5 | NM_001167830.1 | 5 | 1883 | NP_001161302.1 |
| NM_016040.4 | 4 | 1834 | NP_057124.3 | |
| NR_030761.1 | 5 | 1905 | ||
| AK125882.1 | 5 | 1605 | ||
| AL080084.1 | 1 | 381 | ||
| AY359039.1 | 4 | 1492 | AAQ89398.1 | |
| BC016556.1 | 4 | 1499 | AAH16556.1 | |
| BC070051.1 | 4 | 1834 | AAH70051.1 | |
| CR936808.1 | 5 | 1633 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830625 | Chr.1:93100821 - 93269689 on Build GRCh38 | Loss |
|
| nsv1008602 | Chr.1:92711254 - 93175251 on Build GRCh38 | Gain |
|
| nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map