Assay Details
Target Gene Details
Entrez Gene ID: | 8528 |
Gene Name: | D-aspartate oxidase |
Gene Aliases: |
DASOX, DDO-1, DDO-2 |
Location: |
Chr.6:110392180-110415550 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDO | NM_003649.2 | 5 | 1447 | NP_003640.2 |
| NM_004032.2 | 4 | 1270 | NP_004023.2 | |
| BC032786.1 | 5 | 1447 | AAH32786.3 |
More Information
Set Membership: |
|

Genomic Map