Assay Details
Target Gene Details
Entrez Gene ID: | 79980 |
Gene Name: | DSN1 homolog, MIS12 kinetochore complex component |
Gene Aliases: |
C20orf172, KNL3, MIS13, dJ469A13.2, hKNL-3 |
Location: |
Chr.20:36751791-36773827 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSN1 | NM_001145315.1 | NP_001138787.1 | ||
| NM_001145316.1 | NP_001138788.1 | |||
| NM_001145317.1 | NP_001138789.1 | |||
| NM_001145318.1 | NP_001138790.1 | |||
| NM_024918.3 | NP_079194.3 | |||
| XM_006723876.2 | XP_006723939.1 | |||
| XM_017028070.1 | XP_016883559.1 | |||
| XM_017028071.1 | XP_016883560.1 | |||
| XM_017028072.1 | XP_016883561.1 | |||
| AK023408.1 | BAB14564.1 | |||
| AK093031.1 | BAC04024.1 | |||
| AK290738.1 | ||||
| AK301671.1 | ||||
| AK301840.1 | ||||
| BC026011.1 | AAH26011.1 | |||
| BC058899.1 | AAH58899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map