Assay Details
Target Gene Details
Entrez Gene ID: | 9221 |
Gene Name: | nucleolar and coiled-body phosphoprotein 1 |
Gene Aliases: |
NOPP130, NOPP140, NS5ATP13, P130 |
Location: |
Chr.10:102152176-102163871 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOLC1 | NM_001284388.1 | NP_001271317.1 | ||
| NM_001284389.1 | NP_001271318.1 | |||
| NM_004741.4 | NP_004732.2 | |||
| XM_005270273.1 | XP_005270330.1 | |||
| AK056755.1 | ||||
| AK225101.1 | ||||
| AK314363.1 | ||||
| AY820769.1 | AAV67777.1 | |||
| BC001883.1 | AAH01883.1 | |||
| BC006769.2 | AAH06769.1 | |||
| BM473772.1 | ||||
| D21262.1 | BAA04803.1 | |||
| Z34289.1 | CAA84063.1 |
More Information
Set Membership: |
|

Genomic Map