Assay Details
Target Gene Details
Entrez Gene ID: | 10712 |
Gene Name: | family with sequence similarity 189 member B |
Gene Aliases: |
C1orf2, COTE1 |
Location: |
Chr.1:155247205-155255483 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM189B | NM_001267608.1 | NP_001254537.1 | ||
| NM_006589.2 | NP_006580.2 | |||
| NM_198264.1 | NP_937995.1 | |||
| XM_005244845.1 | XP_005244902.1 | |||
| AF070550.1 | AAC28638.1 | |||
| AL353941.1 | ||||
| BC006493.2 | AAH06493.1 | |||
| BC007444.2 | AAH07444.1 | |||
| BC008854.2 | AAH08854.1 | |||
| BC010471.1 | AAH10471.1 | |||
| BC017262.2 | AAH17262.1 | |||
| BC040136.1 | AAH40136.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831636 | Chr.1:155249432 - 155475073 on Build GRCh38 | Loss |
|
| nsv470741 | Chr.1:155142784 - 155260340 on Build GRCh38 | Loss |
|
| esv28935 | Chr.1:155210698 - 155251452 on Build GRCh38 | Gain |
|
| nsv1011773 | Chr.1:155219357 - 155265030 on Build GRCh38 | Gain |
|
| dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map