Assay Details
Target Gene Details
Entrez Gene ID: | 7750 |
Gene Name: | zinc finger MYM-type containing 2 |
Gene Aliases: |
FIM, MYM, RAMP, SCLL, ZNF198 |
Location: |
Chr.13:19957412-20091845 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMYM2 | NM_001190964.2 | 6 | 1396 | NP_001177893.1 |
| NM_001190965.2 | 5 | 1411 | NP_001177894.1 | |
| NM_003453.4 | 6 | 1504 | NP_003444.1 | |
| NM_197968.3 | 5 | 1457 | NP_932072.1 | |
| XM_005266520.3 | 5 | 1416 | XP_005266577.1 | |
| XM_011535216.2 | 6 | 1086 | XP_011533518.1 | |
| XM_011535222.1 | 5 | 1347 | XP_011533524.1 | |
| XM_017020728.1 | 5 | 1347 | XP_016876217.1 | |
| XM_017020729.1 | 5 | 1322 | XP_016876218.1 | |
| XM_017020731.1 | 6 | 1139 | XP_016876220.1 | |
| XM_017020732.1 | 6 | 1161 | XP_016876221.1 | |
| XM_017020733.1 | 7 | 1188 | XP_016876222.1 | |
| XM_017020734.1 | 7 | 1174 | XP_016876223.1 | |
| XM_017020735.1 | 6 | 1138 | XP_016876224.1 | |
| XM_017020736.1 | 6 | 1310 | XP_016876225.1 | |
| XM_017020737.1 | 6 | 1085 | XP_016876226.1 | |
| XM_017020738.1 | 5 | 1400 | XP_016876227.1 | |
| XM_017020739.1 | 5 | 1347 | XP_016876228.1 | |
| AF035374.1 | 3 | 702 | AAB88464.1 | |
| AF060181.1 | 3 | 754 | AAC23591.1 | |
| AJ224901.1 | 6 | 1390 | CAA12204.1 | |
| AK226118.1 | 5 | 1392 | ||
| AK289741.1 | 5 | 1382 | ||
| AK302917.1 | ||||
| AL136621.1 | 6 | 1190 | CAB66556.2 | |
| BC036372.1 | 5 | 1457 | AAH36372.1 | |
| BX647944.1 | 5 | 1411 | CAH56193.1 | |
| Y13472.1 | 6 | 1396 | CAA73875.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832557 | Chr.13:19910435 - 20081915 on Build GRCh38 | Gain |
|
| nsv561215 | Chr.13:19871303 - 20076371 on Build GRCh38 | Gain |
|
| esv2761244 | Chr.13:19389134 - 20043200 on Build GRCh38 | Gain |
|
| nsv1046946 | Chr.13:19730301 - 20507454 on Build GRCh38 | Gain |
|
| esv3892297 | Chr.13:19803308 - 20022425 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115819790] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map