Assay Details
Target Gene Details
Entrez Gene ID: | 50700 |
Gene Name: | retinol dehydrogenase 8 (all-trans) |
Gene Aliases: |
PRRDH, SDR28C2 |
Location: |
Chr.19:10013249-10022278 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RDH8 | NM_015725.2 | NP_056540.2 | ||
| AF229845.1 | AAF63160.1 | |||
| AK024022.1 | BAB14782.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833742 | Chr.19:9872203 - 10021301 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map