Assay Details
Target Gene Details
Entrez Gene ID: | 9338 |
Gene Name: | transcription elongation factor A like 1 |
Gene Aliases: |
SIIR, WEX9, p21, pp21 |
Location: |
Chr.X:103628720-103630953 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCEAL1 | NM_001006640.1 | NP_001006641.1 | ||
| BU602495.1 |
More Information
Set Membership: |
|

Genomic Map