Assay Details
Target Gene Details
Entrez Gene ID: | 91050 |
Gene Name: | coiled-coil domain containing 149 |
Gene Aliases: |
- |
Location: |
Chr.4:24806117-24980204 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC149 | NM_001130726.2 | NP_001124198.1 | ||
| NM_173463.4 | NP_775734.2 | |||
| XM_005248210.3 | XP_005248267.1 | |||
| XM_011513906.2 | XP_011512208.1 | |||
| XM_011513907.2 | XP_011512209.1 | |||
| XM_011513908.1 | XP_011512210.1 | |||
| XM_017008827.1 | XP_016864316.1 | |||
| XM_017008828.1 | XP_016864317.1 | |||
| AK125353.1 | BAC86142.1 | |||
| AK296454.1 | ||||
| AK302905.1 | ||||
| AL834257.2 | 9 | 4749 | CAD38932.1 | |
| BC057761.1 | AAH57761.1 | |||
| BC067735.1 | AAH67735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829887 | Chr.4:24771923 - 25000466 on Build GRCh38 | Gain+Loss |
|

Genomic Map