Assay Details
Target Gene Details
Entrez Gene ID: | 4189 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B9 |
Gene Aliases: |
ERdj4, MDG-1, MDG1, MST049, MSTP049 |
Location: |
Chr.7:108569745-108574850 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB9 | NM_012328.2 | 3 | 2163 | NP_036460.1 |
| AB026908.1 | 3 | 1996 | BAA84703.1 | |
| AK092204.1 | 4 | 2021 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3572052 | Chr.7:108554229 - 108651992 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map