Assay Details
Target Gene Details
Entrez Gene ID: | 1060 |
Gene Name: | centromere protein C |
Gene Aliases: |
CENP-C, CENPC1, MIF2, hcp-4 |
Location: |
Chr.4:67468762-67545601 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CENPC | NM_001812.2 | NP_001803.2 | ||
| XM_006714064.2 | XP_006714127.1 | |||
| AK308214.1 | ||||
| AK309437.1 | ||||
| BC030695.1 | AAH30695.1 | |||
| BC041117.1 | AAH41117.1 | |||
| BX648166.1 | ||||
| M95724.1 | AAA51974.1 |
More Information
Set Membership: |
|

Genomic Map