Assay Details
Target Gene Details
Entrez Gene ID: | 55170 |
Gene Name: | protein arginine methyltransferase 6 |
Gene Aliases: |
HRMT1L6 |
Location: |
Chr.1:107056645-107059294 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRMT6 | NM_018137.2 | 1 | 2024 | NP_060607.2 |
| AK001421.1 | 1 | 1882 | BAA91681.1 | |
| AK297898.1 | ||||
| AY043278.1 | 1 | 1953 | AAK85733.1 | |
| BC002729.2 | 1 | 1845 | ||
| BC063446.1 | 1 | 1825 | ||
| BC073866.1 | 1 | 1972 | AAH73866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758957 | Chr.1:106941006 - 107298261 on Build GRCh38 | Loss |
|
| nsv482070 | Chr.1:107056787 - 107059292 on Build GRCh38 | Gain |
|
| nsv547330 | Chr.1:107006123 - 107375697 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map