Assay Details
Target Gene Details
Entrez Gene ID: | 93650 |
Gene Name: | acid phosphatase, testicular |
Gene Aliases: |
- |
Location: |
Chr.19:50790415-50795224 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACPT | NM_033068.2 | NP_149059.1 |
More Information
Set Membership: |
|

Genomic Map