Assay Details
Target Gene Details
Entrez Gene ID: | 84056 |
Gene Name: | katanin catalytic subunit A1 like 1 |
Gene Aliases: |
- |
Location: |
Chr.13:30202626-30307500 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KATNAL1 | NM_001014380.2 | 11 | 4678 | NP_001014402.1 |
| NM_032116.4 | 11 | 4720 | NP_115492.1 | |
| XM_005266574.2 | 11 | 4862 | XP_005266631.1 | |
| XM_017020791.1 | 10 | 4564 | XP_016876280.1 | |
| XM_017020792.1 | 10 | 4585 | XP_016876281.1 | |
| XM_017020793.1 | 8 | 4894 | XP_016876282.1 | |
| BX647147.1 | 1 | 1418 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631667 | Chr.13:30146805 - 30231044 on Build GRCh38 | Gain |
|
| nsv832572 | Chr.13:30075868 - 30261119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map