Assay Details
Target Gene Details
Entrez Gene ID: | 163351 |
Gene Name: | guanylate binding protein family member 6 |
Gene Aliases: |
- |
Location: |
Chr.1:89363877-89388160 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GBP6 | NM_001320257.1 | 9 | 1673 | NP_001307186.1 |
| NM_198460.2 | 11 | 2014 | NP_940862.2 | |
| XM_011540835.2 | 11 | 1843 | XP_011539137.1 | |
| AI199897.1 | ||||
| AK131329.1 | 6 | 896 | BAD18489.1 | |
| AK131356.1 | 11 | 1998 | BAD18509.1 | |
| AK290303.1 | 11 | 1834 | ||
| AK299408.1 | 10 | 1649 | ||
| AK299466.1 | 9 | 1490 | ||
| AK303987.1 | 11 | 1758 | ||
| AK315834.1 | 11 | 1832 | ||
| BC131713.1 | 11 | 1792 | ||
| BX537949.1 | 11 | 1833 | CAD97917.1 | |
| BX647907.1 | 11 | 1831 |
More Information
Additional Information:
For this assay, SNP(s) [rs76159539] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map